Static Pen, Sartorius
Supplier: Sartorius
YSTP01
76413-056EA 2644.07 USD
76413-056
Static Pen, Sartorius
Static Control Monitors
This pen unit is used to neutralize static electricity on samples or objects to be weighed down to a level of 30V.
The pen features a micro switch to activate the function, main 100 to 230V with 0.1W, 50/60 Hz.