Ionizer Blower, Sartorius
Supplier: Sartorius
YIB01-0UR
76413-010EA 2506.02 USD
76413-010
Ionizer Blower, Sartorius
Ionizers
Ionizing blower for neutralize static electricity within a few seconds, by 'shooting' ions at the material to be weighed. This eliminates the need for increasing of the humidity which can be complicated, expensive, and often ineffective.