Printer for Premium Balances, Sartorius
Supplier: Sartorius
YDP10-0CE
76412-968EA 1454.76 USD
76412-968
Printer for Premium Balances, Sartorius
Printers Document Printers
Data print, for EC stamp approval, with GLP and GMP function for all balances and moisture analyzers.
Metrological department, the PTB. It features an alphanumeric dot matrix print head (safety print head with a 5×8 matrix) as well as the ASCII-chart and application specific symbols for printing weight data, and a touch-control keypad for easy operation.