Calibration Weights, OIML, Class E2, Proof Line, Sartorius
Supplier: Sartorius
YCW412-02
76412-934EA 424.13 USD
76412-934
Calibration Weights, OIML, Class E2, Proof Line, Sartorius
Weights Calibration Weights
Sartorius Proof Line weights are high quality working standards for everyday use. These weights are made of high quality material with polished or precision lathed surfaces.
  • Non-magnetizable
  • Suitable for use in cleanrooms
  • 1 g to 10 Kg Knob weights: Stainless steel
OIML class E2 milligram weights are designed as leaf weights.
Certifications: Comply in all respects with the requirements on shape, material and markings specified by the OIML R111-1 International Recommendation.