Infrared Moisture Analyzer, MA37, Sartorius
Supplier: Sartorius
MA37-1US
76412-336EA 4948.54 USD
76412-336
Infrared Moisture Analyzer, MA37, Sartorius
Analyzers Moisture Analyzers
Compact moisture analyzer giving fast repeatable results with an improved user interface.
  • Convenient process tracking with status light indicator
  • Rapid heat up with powerful heating elements reduces overall process time
  • BetterClean design enables the user to remove the sample chamber plate and heating module and clean in a dishwasher
  • Mini-USB and automatic printer recognition for data communication
Suitable for routine daily tasks and designed to improve the overall operational efficiency from parameter set up through the heating process to final cleaning.