Infrared Moisture Analyzer, MA160, Sartorius
Supplier: Sartorius
MA160-1US
76412-330EA 6371.07 USD
76412-330
Infrared Moisture Analyzer, MA160, Sartorius
Analyzers Moisture Analyzers
Easy to use, efficient, compact moisture analyzers to give fast repeatable results with an improved user interface.
  • Convenient process tracking with status light indicator
  • Rapid heat up with powerful heating elements reduces overall process time
  • BetterClean design enables the user to remove the sample chamber plate and heating module and clean in a dishwasher
  • Easy to operate with touch screen and intuitive user interface
  • Mini-USB and automatic printer recognition for data communication
Designed for routine work and method development for the QC laboratory that needs fast, highly repeatable moisture results, management of several methods for different products and transfer of methods between instruments.
Ensures easy method development and management, as well as simple sample handling and instrument performance testing. The Method Assistant function enables the user to easily develop new methods. Three simple steps are all it takes to develop a new method in less than 45 minutes.