Manual Slim Pen Cutter With Safety Blade, Slice®
Supplier: SLICE, INC.
10476
76411-188CS 254.68 USD
76411-188
Manual Slim Pen Cutter With Safety Blade, Slice®
Knives
The Slice® Manual Slim Pen Cutter uses the same finger-friendly® blade as our original pen cutters, offering the same safety benefits. Slice’s proprietary grind—applied to advanced ceramics—creates a unique edge that’s safe to the touch and lasts up to 11 times longer than comparable metal blades. The slim handle style allows maneuverability in tight spaces and features a side slider for easier manipulation with the thumb. As with all Slice tools, it can be used ambidextrously. Made from durable glass-filled nylon, the Manual Slim Pen Cutter offers two different cutting depths to suit a variety of cutting tasks.
  • Manual retraction with two cutting positions
  • Ambidextrous design; reverse blade orientation for lefties
  • Finger-friendly® blade cuts effectively
  • Reduces injuries; lowers costs
  • Non-sparking, non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
  • Blade edge lasts up to 11.2× longer than metal
At Slice®, we’ve taken our popular pen cutters and added a new variation specifically for working at awkward angles and in tight spaces. These slimmer, more streamlined handles are made from the same sturdy glass-filled nylon material and are compatible with the same 75998-088 (rounded-tip) and 75998-090 (pointed-tip) safety blades as our original pen cutters. Switch the blade orientation for lefties by using our no-tool blade change. Keep your cutter close at hand with its integrated lanyard hole.
It features manual locking retraction with two different cutting depths. Safety is our number one goal: we manufacture proprietary finger-friendly® safety blades that are safe to the touch and cut effectively. Our blades are non-magnetic, non-sparking, oil-free and never rust. They have been proven in third-party tests to last up to 11 times longer than comparable metal blades.
Accessories information: Includes 75998-088 blade. These 100 percent zirconium oxide blades retain their edge 11 times longer than steel and feature our proprietary finger-friendly® grind. Slice ceramic blades require no oil coating or special maintenance; they're non-sparking, non-magnetic, and never rust. The rounded tip offers extra protection against accidental puncture injuries. When you need a fresh edge, simply reverse the double-sided blade.