Auto-Retractable Slim Pen Cutter With Safety Blade, Slice®
Supplier: SLICE, INC.
10475
76411-186CS 236.15 USD
76411-186
Auto-Retractable Slim Pen Cutter With Safety Blade, Slice®
Knives
Spring-loaded automatic blade retraction is just one of the features that makes the Slice® Auto-Retractable Slim Pen Cutter a safer choice. Slice’s proprietary grind—applied to advanced ceramics—creates the only finger-friendly® blade on the market. In addition, our blades never rust and last up to 11 times longer than comparable metal blades. This pen cutter’s durable glass-filled nylon handle has a slim design that’s ideal for working in tight spaces. As with all Slice tools, it can be configured for either right-handed or left-handed use and requires no tools to change the blade.
  • Automatic spring-loaded retraction for added safety
  • Ambidextrous design; reverse blade orientation for lefties
  • Finger-friendly® blade cuts effectively
  • Reduces injuries; lowers costs
  • Non-sparking, non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
  • Oil- and lubricant-free blade
Spring-loaded automatic blade retraction is just one of the features that makes the Slice® Auto-Retractable Slim Pen Cutter a safer choice. Slice’s proprietary grind—applied to advanced ceramics—creates the only finger-friendly® blade on the market. In addition, our blades never rust and last up to 11 times longer than comparable metal blades. This pen cutter’s durable glass-filled nylon handle has a slim design that’s ideal for working in tight spaces. As with all Slice tools, it can be configured for either right-handed or left-handed use and requires no tools to change the blade. Keep your Auto-Retractable Slim Pen Cutter close by with its convenient integrated lanyard hole.
Accessories information: Includes 75998-088 blade. These 100 percent zirconium oxide blades retain their edge 11 times longer than steel and feature our proprietary finger-friendly® grind. Slice ceramic blades require no oil coating or special maintenance; they're non-sparking, non-magnetic, and never rust. The rounded tip offers extra protection against accidental puncture injuries. When you need a fresh edge, simply reverse the double-sided blade.