Auto-Retractable Small Scraper, Slice®
Supplier: SLICE, INC.
10590
76411-182CS 236.15 USD
76411-182
Auto-Retractable Small Scraper, Slice®
Scrapers
This small scraper features spring-loaded auto-retraction so you cannot accidentally leave the blade exposed. To avoid excess strain, Slice® invented SureScrape™, which absorbs kickback force, so you don’t have to. An installed safety blade features a proprietary finger-friendly® edge, which scrapes effectively and is safe to the touch.
  • Ambidextrous design
  • Non-sparking, non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
  • Oil- and lubricant-free blade
  • 100% zirconium oxide blade
  • Blade edge lasts up to 11.2× longer than metal
  • No-tool blade change
This small scraper features spring-loaded auto-retraction so you cannot accidentally leave the blade exposed. To avoid excess strain on thumb and wrist joints from scraping resistance, Slice® invented SureScrape™, which absorbs kickback force, so you don’t have to. No other manufacturer offers a comparable ergonomic advantage. The 10590 ships with a rounded-tip 10524 blade and is compatible with 10523 (serrated) and 10525 (pointed-tip) Slice blades. All Slice safety blades feature a proprietary finger-friendly® edge, which scrapes effectively and is safe to the touch. Blades are non-magnetic, non-conductive, and never rust.