Manual Mini Scraper With Safety Blade, Slice®
Supplier: SLICE, INC.
10594
76411-176CS 236.15 USD
76411-176
Manual Mini Scraper With Safety Blade, Slice®
Knives
This smaller version of Slice’s original manual scraper uses a narrower, thinner blade and is intended for tight spaces and small surfaces.
  • Durable nylon handle with textured slider
  • Manual retraction with 1 scraping position
  • Ambidextrous design
  • Finger-friendly® blade scrapes effectively
  • Reduces injuries, lowers costs
  • Non-sparking, non-conductive, non-magnetic blade
  • Chemically inert blade never rusts
While most people prefer a straight-edge blade (like Slice® 75998-096 or 76314-922 blades), this handy tool is also compatible with the serrated 76314-924 blade. All blade options are finger-friendly® and last up to 11× longer than metal blades. In addition, these 100 percent zirconium oxide blades are non-sparking, non-conductive, non-magnetic, and will never rust. The durable glass-filled nylon handle features a textured slider for comfort. Blade locks in place when in use and retracts fully afterwards for safety.
Accessories information: Includes 75998-096. Slice® Utility Blades (Rounded Tip) are ideal for handheld applications, such as trimming flash off injection-molded plastic, or for use in third-party handles. Slice developed these blades before creating our own line of utility knives. We recommend that customers try Slice utility knives, but if you need a larger blade for hand-held applications. It is made with advanced ceramics, which are non-sparking, non-magnetic, chemically inert, safe up to 1600 °C, and never rust.