Vivaspin® Endotest, Centrifugal Filtration Units, Sartorius
Supplier: Sartorius
VS15RXETO
76409-040PK 200.18 USD
76409-040
Vivaspin® Endotest, Centrifugal Filtration Units, Sartorius
Filters Centrifugal Filters
For 2 to 15 ml, cellulose triacetate membrane. Disposable ultrafiltration device for endotoxin concentration and removal of substances interfering with the LAL test from liquid samples. The device is a spin column based ultrafiltration unit. It can effectively be used in swing bucket or fixed angle rotors accepting 50 ml conical bottom centrifuge tubes.
  • Minimises hands on time
  • Ideal for concentration of samples for endotoxin free applications