Vivapore® Solvent Absorption Concentrators, Sartorius
Supplier: Sartorius
VP0503 VP2002 VP2003
76408-846PK 292.42 USD
76408-846 76408-848 76408-850 76408-852 76408-854 76408-856
Vivapore® Solvent Absorption Concentrators, Sartorius
Concentrators
Ideal for general purpose laboratory concentration or purification prior to further analysis, Vivapore® is particularly suited for labile solutions that can denature with alternative shear or pressure inducing methods or that require processing in a cold room environment.
  • Sample concentration without additional equipment
  • Separate chambers prevents cross contamination
  • SAN reservoir material
With no need for additional equipment, pressure or vacuum, solvent absorption is the most economic and user friendly concentration technique available to the clinician and research scientist.
Just fill the unit with the solution to be concentrated, wait for the desired concentration level to be achieved and then pipette the concentrated sample from the bottom of the reservoir.
Vivapore 5: 1 - 5 ml, 20 cm² of membrane surface area
Vivapore 10/20: 2 - 10 ml (up to 20 ml with expansion reservoir in option), 28 cm² of membrane surface area