Vivaclear Centrifuge Filters, Sartorius

Supplier: Sartorius
VK01P042
76408-740PK 437.73 USD
76408-740
Vivaclear Centrifuge Filters, Sartorius
Filters Centrifugal Filters
Vivaclear centrifugal filters are disposable microfiltration devices for the fast and reliable clarification/filtration of biological samples in the range 100 to 500 µl. They can be used in fixed angle rotors suitable for 2.2 ml centrifuge tubes.

  • High-flux polyethersulfone membrane
  • Low hold-up volume (<5 µl) with total length of 43 mm
  • Fast and reproducible performance
  • Rotor: 40 to 45° fixed-angle rotor, 500 µl
  • Filtrate collection tube Ø: 11 mm
  • Body and filtrate collection tube are made up of polypropylene

Applications: Clarification of samples before loading onto Vivapure protein purification spin columns; removal of particles and participates; filtration of plasma and serum; filtration of cells or cell debris.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR