Corepoint Scientific™ Manual Defrost Hydrocarbon Laboratory and Medical Freezer, Horizon Scientific
Supplier: Horizon Scientific
NSPF201WWW/0MHC
76407-446EA 3764.98 USD
76407-446
Corepoint Scientific™ Manual Defrost Hydrocarbon Laboratory and Medical Freezer, Horizon Scientific
Freezers Laboratory / General Purpose Freezers
These White Diamond laboratory freezers provide professional cooling for scientific and clinical applications.
  • Microprocessor temperature control
  • Integrated temperature alarms
  • Forced draft air circulation
  • Probe access
  • Keyed door locks
  • Manual defrost
  • Energy Star Certified
  • Natural hydrocarbon refrigerants
  • ENERGY STAR certified*
These combination units come equipped with forced draft circulation, integrated alarms, and precise temperature monitoring through microprocessor control. All units run on natural, hydrocarbon refrigerant for environmental health and energy efficiency.
Certifications: *Select models ENERGY STAR certified: 76407-446
Ordering information: All units come with a two year parts and labor warranty, plus an additional three year compressor parts warranty.