Luna® Cell Ready One-Step RT-qPCR Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E3030S
76407-356EA 1164 USD
76407-356
Luna® Cell Ready One-Step RT-qPCR Kit, New England Biolabs
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
The Luna® Cell Ready One-Step RT-qPCR Kit provides all the necessary components for direct, dye-based RNA detection and quantitation, bypassing the need for RNA extraction and purification.
  • Contains reagents for 100 lysis preps and 500 one-step RT-qPCR reactions (dye)
  • Effective cell lysis preparation from 10 to 100000 cells across numerous cell lines
  • Coordinated cell lysis, RNA release, and genomic DNA removal in a fast 15-minute protocol
  • Luna WarmStart RT paired with Hot Start Taq for increased thermostability and room temperature setup
Coordinating the actions of DNase I and the Luna Cell Ready Protease, the Luna Cell Ready Lysis Module offers a simple alternative workflow resulting in effective cell lysis, RNA release, and genomic DNA removal simultaneously in a 15-minute protocol. The Lysis Module includes a unique Luna Cell Ready RNA Protection Reagent that maintains RNA integrity during cell lysis. The lysis capacity spans 10 to 100000 cells in a 50 µl lysis reaction. Up to 2 µl of lysate (equivalent to RNA from 0.2 to 4000 cells) can be transferred into 20 µl downstream RT-qPCR reactions.
Delivery information: The Luna Cell Ready One-Step RT-qPCR Kit is composed of:
1) The Luna Cell Ready Lysis Module
2) The Luna Universal One-Step RT-qPCR Kit.