VWR® Incubated Shakers
Supplier: VWR International
H1010-G-VWR
76407-112EA 8895.97 USD
76407-112
VWR® Incubated Shakers
Shakers and Mixers Shaking Incubators
High capacity benchtop incubated shaker that accepts up to 9×1 or 5×2 L flasks.
  • LED display for controlling over time, temperature and mixing speed
  • Unique platform accepts magnetic clamps for easy attachment of flasks and bottles
The VWR® incubated shakers are compact orbital shakers with precision temperature control.
Other models in the series are available offering a variety of temperature range capabilities and chamber sizes.
Certifications: CE marked.
Ordering information: Supplied without accessory clamps. A wide range of accessories are available to order separately.
Supplied with a standard non-skid, silicone mat that is suitable for low speed shaking of tissue culture flasks, Petri dishes and staining trays. For high speed shaking, an optional universal/magnetic platform is available with clamp accessories for a variety of flasks, tubes and other common vessels.