CloneSelect™ Imager, Molecular Devices
Supplier: Molecular Devices
CLONESELECTIMG
76404-532EA 119753.18 USD
76404-532
CloneSelect™ Imager, Molecular Devices
Colony Pickers
Overcome the challenges associated with conventional cell confluence techniques with the CloneSelect Imager system - producing objective, quantitative, and consistent results in less time.
  • Assess cell confluence objectively and quantitatively
  • Streamline workflow: image, analyze, report
  • Image every well anytime to track colony formation and verify monoclonality in every well at any time point
  • Verify monoclonality and image image every well, at any time point in time
  • Includes internal barcode reader for data tracking of each plate
  • Rapid, automated, label-free imaging for monitoring cell growth and assurance of monoclonality
The CloneSelect™ Imager is a high-throughput automated solution for imaging and analyzing mammalian cells. Tracking the formation of a colony from a single cell is effortless as barcoded plates are tracked over time. Automated acquisition and analysis provide accurate, objective, and consistent results. Detailed imaging and comprehensive data analysis enable better decision making. With the ability to image a 96-well plate in 90 seconds, the imager will increase throughput and productivity.
Plate Type - Range of 6-, 12-, 24-, 96- and 384-well SBS microplates.
Monoclonality report feature streamlines the creation of supporting documentation for regulatory agencies.
Certifications: CE, ISO9001:2008 certified