HistoCore AUTOCUT Automated Rotary Microtome
Supplier: Leica Biosystems MS
149AUTO00C1
76404-254EA 39026.32 USD
76404-254
HistoCore AUTOCUT Automated Rotary Microtome
Microtomes and Microtome Assemblies Microtomes
A microtome unlike any other.
  • Every microtomist can use their prefrerred sectioning mode
  • Automated: motorized push-button sectioning
  • Semi-automated: push-button specimen advance and retract
  • Manual: full control for those who love the art of microtomy
  • Maximize safety in the lab: The AUTOCUT improves on past automated models with an automatic electronic brake to minimize the risk of injury during operation
  • Shorten cleaning time from minutes to seconds with the Antistatic Waste Tray
HistoCore AUTOCUT allows users to select among automated, semi auto-automated, or manual sectioning modes based their personal preferences.
As soon as you pause the automated sectioning function, the Electronic Brake kicks in immediately to return and hold the specimen in the safest position away from the blade. This helps to minimize injury to the operator when handling the specimen.
With the antistatic waste tray, AUTOCUT helps you keep your workplace clean, efficient, and debris-free. The tray is made from antistatic material which prevents paraffin waste from sticking to the walls. It provides unmatched efficiency by reducing your cleaning time from minutes to seconds.
More surface area on top of the microtome, so you can store the tools you need for your job – brushes, blades, wipes, and more.
Delivery information: Configuration 1 model features a rotary microtome with automated, semi-automated, and manual modes, automatic electronic brake. Includes antistatic section waste tray, top tray, precision orientation with quick clamping system for specimen clamps,
universal cassette clamp, and two-in-one blade holder E for high and low-profile blades with lateral displacement function (includes blade holder base).