Leica HistoCore NANOCUT R Automated Research and Special Applications Rotary Microtome
Supplier: Leica Biosystems MS
14052458261
76404-250EA 52375.76 USD
76404-250
Leica HistoCore NANOCUT R Automated Research and Special Applications Rotary Microtome
Microtomes and Microtome Assemblies Microtomes
When microtomy involves diverse samples in order to discover new breakthroughs in research projects, then NANOCUT R is the optimal solution. From biomedical research to industrial applications, NANOCUT R can cut virtually anything can be put in front of it.
  • Cut sections that can be prepared for analysis in light microscopy and electron microscopy - down to 250 nm (0.25 µm, semi-thin)
  • Operating humidity range of 20 to 80% maximum (non-condensing)
  • Maximum heat emission of 340 J/s
Preserve sample integrity when performing semi-thin sectioning with the special spindle and expanded slow speed adjustment. Precisely stop the sample in the object head's optimal position for photo-capture of the cut sample surface when running 3-dimensional reconstruction programs.
Operating temperature: 18...35 °C