Cryostat, CM1860, Leica
Supplier: Leica Biosystems MS
149491860US
76404-232EA 51387.58 USD
76404-232
Cryostat, CM1860, Leica
Microtomes and Microtome Assemblies Microtomes
This high-throughput cryostat delivers diagnostic confidence by reliably producing quality sections to help provide an accurate diagnosis - even with the most complicated tissue types.
  • CM1860 is a perfect assistant for high-quality, fast and safe sectioning
  • Standard applications in the clinical histopathology laboratory
Staff near the cryostat is protected by the antimicrobial nano-silver coating, silver protect, which provides outstanding safety by reducing exposure to surface pathogens. Silver protect covers the cryostat’s external upper surfaces and constantly protects individuals in the work area by penetrating the membranes of microbes to prevent replication.
The unique premium blade holder safety features combined with the ergonomics of a designated finger guard, blade ejector and palm rest for brush technique users improve safety and comfort while sectioning.