TitroLine® 7500 KF Titrators, SI Analytics
Supplier: YSI
285230350
76403-718EA 7670.01 USD
76403-718
TitroLine® 7500 KF Titrators, SI Analytics
Titrators
The TitroLine® 7500 KF offers additional performance capabilities and is the volumetric generalist for a wide range of use.
Its design includes a high visible full colors display that can be easily viewed from a distance and extreme angles. It is also capable of storing results via USB port with the capability of being exported as either a PDF or CSV format. This system is also equipped with intelligent interchangeable modules.
It is designed for fast, easy and precise usage. It comes equipped with standard methods for different applications (titer determination, blank value…).