Halogen Moisture Analyzer HX204, METTLER TOLEDO®
Supplier: Mettler Toledo
30019875
76403-494EA 16180 USD
76403-494
Halogen Moisture Analyzer HX204, METTLER TOLEDO®
Analyzers Moisture Analyzers
Halogen Moisture Analyzer HX204 for high performance moisture analysis.
  • Sample weight 0.1 g to 200 g
High Performance Moisture Analysis – 0.001 %MC (0.1 mg) readability, user guidance, real-time drying curve, user management, method (300) and result (3000) storage, control charts, 4 drying programs, FACT, automatic lid.
Accurate Results for All Sample Types – Get highly reliable and repeatable moisture results thanks to fast halogen heating and superior weighing performance with high 0.001% MC readability.
Intuitive OneClick™ Operation – The large 7" color touchscreen offers an easy to navigate user interface. The operator is guided graphically step by step through the workflow.
Increased Safety with User Management – Define multiple users with individual rights, each with their own home screen and method shortcuts. This reduces errors and increases efficiency.
Built-in Tests Assure Performance – Integrated routine testing features ensure the instrument is always performing optimally: FACT, SmartCal and tests for the heating and weighing units.
Ordering information: Temperature adjustment kit available for easy test and adjustment by user; contact your Avantor representative for more information.