Epower™ Burkholderia multivorans Derived from ATCC® BAA-247™
Supplier: MICROBIOLOGICS, INC.
01270E7
76402-990EA 480.59 USD
76402-990
Epower™ Burkholderia multivorans Derived from ATCC® BAA-247™
Controls and Calibration Verification Materials
10 pellets of a single microorganism with a predetermined quantitative assay
Strain Characteristics: Quantitative preparations at a concentration of 1.0-9.9E+07 CFU per pellet