Superspher® RP select B, HPLC Columns
Supplier: Avantor
SUPERRPB4125AF SUPERRPB4150AF SUPERRPB4250AF
76402-404EA 1425.88 USD
76402-404 76402-406 76402-408 76402-410
Superspher® RP select B, HPLC Columns
Chromatography Columns
A range of 4 µm materials has been marketed under the tradename Superspher®.
  • Spherical porous silica
  • Base deactivated LiChrospher RP-select B
  • Endcapping options for RP-8 and RP-18
Hichrom high efficiency packed columns.