Superspher® RP8, HPLC Columns
Supplier: HICHROM LIMITED
SUPERRP84250AF
76402-390EA 419.34 USD
76402-390 76402-392 76402-394 76402-396 76402-398 76402-400 76402-402
Superspher® RP8, HPLC Columns
Chromatography Columns
A range of 4 µm materials has been marketed under the trade name Superspher®.
  • Spherical porous silica
  • Base deactivated LiChrospher RP-select B
  • Endcapping options for RP-8 and RP-18
Hichrom high efficiency packed columns.