Superspher® RP18-E, HPLC Columns
Supplier: HICHROM LIMITED
SUPERRP18E412F
76402-354EA 959.66 USD
76402-354 76402-370 76402-372 76402-374 76402-376 76402-378 76402-380 76402-382 76402-384 76402-386
Superspher® RP18-E, HPLC Columns
Chromatography Columns
A range of 4 µm materials has been marketed under the Superspher® tradename in Europe.
  • Spherical porous silica
  • Base deactivated LiChrospher RP-select B
  • Endcapping options for RP-8 and RP-18
  • Hichrom high efficiency packed columns