Avantor® Hichrom, In-Line Filter
Supplier: HICHROM LIMITED
HI-683
76399-690EA 302.6 USD
76399-690
Avantor® Hichrom, In-Line Filter
Filters In-Line Filters
This is a fingertight bioanalytical inline filter, comprising a PEEK holder and a 5 μm titanium PEEK-encased frit.
  • Protects the injector from particulates
  • Compatible with all HPLC systems
  • Replacement 5 μm and 2 μm titanium PEEK-encased frits are available
Placed between the pump and sample injection valve, the in-line filter traps particles released through normal piston seal wear within the pump. It is recommended that the frits are checked frequently and replaced as soon as they contribute to a system back pressure.