Avantor® Hichrom, ValvTool
Supplier: HICHROM LIMITED
HI-199
76399-202EA 97.41 USD
76399-202
Avantor® Hichrom, ValvTool
Wrenches
This Valvtool is a uniquely designed slotted wrench which provides easy access to many hard-to-reach areas.
It enables fittings, sample loops and most other ¹/₄" stainless steel and PEEK HPLC fittings to be more easily tightened or loosened.