Avantor® Hichrom, Pliers for Stainless Steel Tubing
Supplier: Avantor
HI-196
76399-196EA 67.33 USD
76399-196
Avantor® Hichrom, Pliers for Stainless Steel Tubing
Pliers
These pliers are designed for cutting ¹/₁₆" stainless steel tubing and are especially suitable for tubing of 0.010" and 0.020" ID.
  • Leaves smooth and uniform cuts
  • De-burring may be necessary to achieve an optimum finish