Avantor® Hichrom, Tubing Cutter for Polymer Tubing
Supplier: HICHROM LIMITED
HI-191
76399-188EA 71.4 USD
76399-188
Avantor® Hichrom, Tubing Cutter for Polymer Tubing
Cutters
Standard cutter for 1/16" and 1/8" O.D. tubing.
  • Compatible with PEEK and PTFE tubing
  • Flat burr-free cuts
  • Guide holes to ensure precise cutting
Delivery information: Includes five replacement blades.