Avantor® Hichrom, Tubing Sleeves, PEEK, MicroTight
Supplier: HICHROM LIMITED
HI-171
76399-176EA 80.25 USD
76399-176
Avantor® Hichrom, Tubing Sleeves, PEEK, MicroTight
Tubing Chromatography Tubing
These tubing sleeves are made of PEEK.
  • Colour-coded for easy I.D. identification
  • Temperature is stable upto 125 °C
These tubing sleeves are used with MicroTight fittings to connect capillary tubing. The I.Ø of the sleeve should be 0.001 to 0.002" larger than the O.Ø of the capillary tubing.
Pressure of the sleeve is 276 bar (4003 psi) and the O.Ø is 0.635 mm.