Avantor® Hichrom, Fused Silica Tubing
Supplier: HICHROM LIMITED
HI-1097
76399-066EA 24.08 USD
76399-066
Avantor® Hichrom, Fused Silica Tubing
Tubing Chromatography Tubing
The maximum operating temperature is 100 °C.
  • pH range 0 to 10
  • Used for micro and nano scale HPLC and capillary electrophorosis
Manufactured from synthetic fused silica with a polyimide coating.