Avantor® Hichrom PEEKsil™, Tubing
Supplier: Avantor
HI-1083
76399-056PK 290.46 USD
76399-056 76399-354
Avantor® Hichrom PEEKsil™, Tubing
Tubing Chromatography Tubing
PEEKsil™ tubing is mechanically strong and has ideal characteristics for sealing with polymer fittings. It is comprised of high quality fused silica sheathed by PEEK tubing giving it excellent chemical compatibility.