Exsil ODS1, HPLC Columns
Supplier: Avantor
EXODS1-5-250A
76398-638EA 283.76 USD
76398-638 76398-784 76398-786 76398-788 76398-790 76398-792 76398-794 76398-796 76398-798 76398-800 76398-802 76398-804 76398-806
Exsil ODS1, HPLC Columns
Chromatography Columns
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
  • Spherical porous silica
  • Competitively priced
  • Hichrom high efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications.