Exsil C8, HPLC Columns
Supplier: Avantor
EXC8-3-150A
76398-626EA 604.49 USD
76398-626 76398-662 76398-664 76398-666 76398-668 76398-670 76398-672 76398-674 76398-676 76398-678
Exsil C8, HPLC Columns
Chromatography Columns
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
  • Spherical porous silica
  • Competitively priced
  • High efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications. Exsil 80 is a higher surface area, complementary range of materials.