Exsil Silica, HPLC Columns
Supplier: Avantor
EX-5-250A
76398-598EA 592.36 USD
76398-598 76398-600 76398-602 76398-604 76398-606 76398-608 76398-610 76398-612 76398-614 76398-616 76398-618 76398-620 76398-622 76398-624
Exsil Silica, HPLC Columns
Chromatography Columns
Exsil HPLC columns are a useful alternative to the classical spherical HPLC silicas.
  • Spherical porous silica
  • 80 and 100 Å pore sizes
  • Competitively priced
  • Hichrom high efficiency
Exsil 100, the standard material, is supplied in a range of particle sizes and bonded phases to cover a wide range of applications. Exsil 80 is a higher surface area, complementary range of materials.