Avantor® Partisphere C8, HPLC Columns, 5 µm
Supplier: Avantor
4621-0503
76398-278EA 994.82 USD
76398-278
Avantor® Partisphere C8, HPLC Columns, 5 µm
Chromatography Columns
Avantor® Partisphere® was one of the first commercially available spherical silicas and continues to provide reproducible, high efficiency separations.
  • Spherical 5 µm porous silica
  • Unique void sealing cartridge hardware
  • Convenient and easy to use - hand tightened fittings
  • Increased column lifetime
Stainless steel column hardware.