Avantor® Partisphere C18, HPLC Columns, 5 µm
Supplier: Avantor
4621-0502
76398-268EA 1201.27 USD
76398-268 76398-270 76398-276 76398-288
Avantor® Partisphere C18, HPLC Columns, 5 µm
Chromatography Columns
Avantor® Partisphere® was one of the first commercially available spherical silicas and continues to provide reproducible, high efficiency separations.
  • Spherical 5 µm porous silica
  • Unique void sealing cartridge hardware
  • Convenient and easy to use - hand tightened fittings
  • Increased column lifetime
Stainless steel column hardware.