Avantor® Ultrasphere® XL ODS (8), Cartridge Columns
Supplier: Avantor
237521
76398-120EA 910.09 USD
76398-120 76398-124 76398-132
Avantor® Ultrasphere® XL ODS (8), Cartridge Columns
Chromatography Columns
Avantor® Ultrasphere® columns are available in variety of phases with 2 range of particle size.
  • High reproducibility
  • Widely referenced silica
  • Optimal surface coverage for long column lifetime
  • Narrow particle size distribution for high efficiency and improved resolution
Avantor® Ultrasphere® Octyl (3 µm) is available in both standard analytical column hardware and a high efficiency Ultrasphere-XL High Speed cartridge column design, which includes an integral cartridge column holder assembly.
Ordering information: Integral cartridge column holder assembly must be ordered separately.