Vydac 202TP C18-HL, HPLC Columns
Supplier: Avantor
202TP54
75798-288EA 1199.91 USD
75798-288 76398-028 76398-030
Vydac 202TP C18-HL, HPLC Columns
Chromatography Columns
Vydac TP was the first range of wide pore media phases developed in the Vydac range and became the industry standard for peptide and protein separations.
  • Long column lifetime and negligible phase leaching
  • Reliable protein purifications, scalable from analytical to preparative scale
  • Referenced in a large number of patents and publications
  • USP L1
Higher carbon load than 201TP. Developed for the separation of PAHs.
The large pores of the 300 Å TP silica give polypeptide molecules complete access to the interior of the silica pores.
Vydac is a registered trademark of W.R.Grace and Co.