SlideMate™ Pro Microscope Slide Printer, Epredia™
Supplier: Epredia
A83900015
76397-806EA 40407.78 USD
76397-806
SlideMate™ Pro Microscope Slide Printer, Epredia™
Printers Cassette and Slide Printers
Epredia™ SlideMate™ Pro slide printer is a smart solution with flexible configuration options.
  • The SlideMate™ Pro printer features a dual-hopper loading system with intelligent slide selection for efficient performance
  • Avoid wasted time switching between multiple slide types
  • Or maximize throughput by loading both hoppers with matching slides
  • The SlideMate™ Pro has a high throughput: 4 to 5 seconds per slide
Setup is simple. The SlideMate™ Pro software uses a Windows® based driver that easily connects to other computers and laboratory information systems. The SlideMate™ Pro enables sample-tracking solutions including user log-in information, time and date stamp on each slide, and unique transaction IDs.
Accessories information: SlideMate™ Pro Spare Slide Hopper is an optional accessory to be used with the Epredia SlideMate™ Pro thermal slide printer. It provides additional flexibility to quickly change slide type/tab color during printer use.