Avantor® ACE® HILIC-B, HPLC Columns, 5 µm
Supplier: Avantor
HILB-5-2520
76395-678PK 1575.68 USD
76395-678 76395-712 76395-714 76395-716 76395-718 76395-734 76395-736 76395-738 76395-740 76395-760 76395-776 76395-778 76395-780 76395-782 76395-798 76395-800 76395-802 76395-804 76395-808 76395-810 76395-812 76395-814 76395-838 76395-840
Avantor® ACE® HILIC-B, HPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® HILIC-B columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of three proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
  • Proprietary aminopropyl basic character HILIC phase
  • Compatible with all manufacturers HPLC/UHPLC systems
  • Excellent column performance, inertness and robustness
  • Simplified and structured approach to successful HILIC separations
Column hardware: Stainless steel.
Ordering information: Available in a range of particle sizes and column dimensions including UHPLC/HPLC rated hardware formats.