Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, 2 µm
Supplier: Avantor
EXL-1011-0502U EXL-1011-1502U EXL-1011-0202U EXL-1011-0203U EXL-1011-0302U EXL-1011-0303U EXL-1011-0503U EXL-1011-0546U EXL-1011-1002U EXL-1011-1003U EXL-1011-1046U EXL-1011-1202U EXL-1011-1203U EXL-1011-1246U EXL-1011-1503U EXL-1011-1546U EXL-1011-7502U EXL-1011-7503U EXL-1011-7546U
76382-616EA 1521 USD
76382-616 76382-618 76392-542 76392-544 76392-546 76392-548 76392-550 76392-552 76392-556 76392-558 76392-560 76392-562 76392-564 76392-566 76392-568 76392-570 76392-572 76392-574 76392-576 76392-584 76392-586 76392-588 76392-594 76392-596 76392-598
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, 2 µm
Chromatography Columns
Avantor® ACE® Excel® SuperC18™ columns provide high stability across an extended pH range. These columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.
  • Ultra-inert for maximum performance and reproducibility
  • Ultra low bleed for improved LC/MS compatibility
  • Wide range of particle sizes (1.7 to 10 µm) and complementary phases
These columns are made of stainless steel.