Combo Wrist Strap and Footwear Tester with Stand, SCS, Desco Industries
Supplier: Desco
770031
76382-940EA 504 USD
76382-940
Combo Wrist Strap and Footwear Tester with Stand, SCS, Desco Industries
Grounders Wrist Grounders
This Combo Wrist Strap and Footwear Tester designed to test the operator’s path-to-ground, and to determine that the operator’s personnel grounding devices (wrist straps, foot grounders, ESD shoes, smocks) are functioning correctly.
  • Separate test circuits for wrist straps and foot grounders
  • Rocker switch allows operator to quickly test all personal grounding devices
  • Use with single-wire wrist straps
  • Durable, stainless steel test button
  • Powered by 9 V battery or optional power adapter
  • Made in the United States of America
This tester verifies the functionality of an operator’s wrist strap and footwear (footwear test requires foot plate). It determines if an operator’s wrist strap and footwear will function correctly. A rocker switch allows the operator to select a wrist strap or footwear test. A green light indicates that the wrist strap or footwear is passing. Red and yellow lights plus an audible alarm indicate when the wrist strap or footwear is failing. If failure occurs, the tester will also display if the grounding device’s resistance is too low or too high. The Combo Tester may also be used to test smocks or garments that feature a grounding mechanism for operators using a coiled cord connection.
Wrist Strap: 750 kilohms and 10 megohms
Footwear: 750 kilohms and 100 megohms
Certifications: NIST calibrated with certificate included.
Delivery information: Combo tester is supplied with stand and foot plate.