Hughes Portable, Self-Contained 16 Gallon Gravity-Fed Eyewash Station, Justrite®
Supplier: Justrite
16GFEW
76382-754EA 512.68 USD
76382-754
Hughes Portable, Self-Contained 16 Gallon Gravity-Fed Eyewash Station, Justrite®
First Aid Supplies Eyewash Stations and Eyewash Bottles
The hughes portable, self-contained 16 gal. gravity-fed eyewash station requires no plumbing, and is suitable for use wherever continuous water is not available.
  • Station provides a controlled 1.5 L/min. (0.4 gal./min.) flow for 15 minutes
  • Portable design makes it easy to move and relocate
  • Station can be wall mounted or used on a bench or table top
The gravity-fed eyewash station supplies a continuous, controlled flow that exceeds the ANSI Z358.1 standard of 1.5 LPM (0.4 GPM) for 15 minutes, providing a reliable safety solution no matter where it isMore Product Information