Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
ACE-121-0103GD ACE-121-0502 ACE-121-0546 ACE-121-1546 ACE-121-2546 ACE-121-0101GD ACE-121-0102GD ACE-121-0104GD ACE-121-0110GD ACE-121-0346 ACE-121-0503 ACE-121-0504 ACE-121-12003 ACE-121-1203 ACE-121-1204 ACE-121-1220 ACE-121-1246 ACE-121-3346 ACE-121-7502 ACE-121-7546 ACE-121-3002 ACE-121-3004 ACE-121-3039 ACE-121-3046 ACE-121-3504 ACE-121-3546 ACE-121-0120GD ACE-121-0202 ACE-121-0203 ACE-121-0246 ACE-121-0302 ACE-1211000075 ACE-121-1002 ACE-121-1003 ACE-121-1004 ACE-121-1008 ACE-121-1008E ACE-121-1010 ACE-121-1020 ACE-121-1046 ACE-121-1502 ACE-121-1503 ACE-121-1504 ACE-121-1508 ACE-121-1510 ACE-121-1520 ACE-121-1539 ACE-121-2002 ACE-121-2004 ACE-121-2046 ACE-121-2502 ACE-121-2503 ACE-121-2504 ACE-121-2508 ACE-121-2510 ACE-121-2520 ACE-121-2530 ACE-121-2539 ACE1212546MVK ACE-121-2546P
76382-572PK 462.26 USD
76382-572 76382-574 76382-576 76382-578 76382-580 76385-748 76385-752 76385-758 76385-760 76385-898 76385-912 76385-914 76385-916 76385-918 76385-922 76385-924 76385-926 76385-928 76385-930 76385-932 76385-934 76385-936 76386-080 76386-082 76386-084 76386-086 76386-088 76386-090 76386-092 76386-094 76386-096 76386-102 76386-104 76386-106 76386-456 76386-458 76386-602 76386-604 76386-606 76386-608 76386-610 76386-612 76386-614 76386-616 76386-622 76386-624 76386-626 76386-336 76386-338 76386-340 76386-342 76386-344 76386-348 76386-350 76386-360 76386-362 76386-372 76386-374 76386-460 76386-462 76386-464 76386-466 76386-468 76385-702 76385-780 76385-822 76385-824 76385-826 76385-870 76385-876 76385-878 76385-882 76385-886 76385-888 76385-892 76385-938 76385-966 76385-968 76385-970 76385-972 76385-974 76385-976 76385-978 76385-980 76385-982 76385-984 76385-988 76385-990 76385-992 76386-002 76386-004 76386-014 76386-126 76386-128 76386-130 76386-132 76386-134 76386-136 76386-138 76386-140 76386-142 76386-144 76386-148 76386-150 76386-152 76386-154 76386-156 76386-158 76386-176 76386-178 76386-198 76386-286 76386-288 76386-290 76386-292 76386-294 76386-296 76386-298 76386-300 76386-302 76386-304 76386-306 76386-308 76386-310 76386-312 76386-314 76386-316 76386-318
Avantor® ACE® C18, HPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.