Argos Technologies® Omega® Multichannel Electronic Pipettors, Cole Parmer
Supplier: Antyila Scientific
24501-30 24501-31 24501-32 24501-33 24501-34 24501-35 24501-36 24501-37 24501-38 24501-39
76379-906EA 882.51 USD
76379-906 76379-908 76379-910 76379-912 76379-914 76379-916 76379-918 76379-920 76379-922 76379-924
Argos Technologies® Omega® Multichannel Electronic Pipettors, Cole Parmer
Pipettes Micropipettes
Ergonomic design for effortless dispensing.
  • Five essential modes: forward, reverse, multi, mixing and diluting
  • Pipetting convenience: 360° rotational manifold
  • Auto-sensing magnetic piston for controlled pipetting of individual channels
  • Specially designed tip cones for leak-free operation
  • Drop of the pipette is color-coded by volume for easy identification
  • Smooth and light-loading ejection of tips
Designed to operate up to five different modes of pipetting, these pipettes also feature the capability of storing up to three of your more frequently used modes. Another feature is a display panel for ease of use. They have a smart power saver, which activates a sleep mode enhancing its battery life.
Its battery charges by either a charging stand or USB power cord and does so in two hours, lasting up to five hours of continuous pipetting.