D/E Neutralizing Broth
Supplier: HARDY DIAGNOSTICS
U332
76377-556PK 221.48 USD
76377-556
D/E Neutralizing Broth
Media for Microbiology Prepared Media for Microbiology
D/E Neutralizing Broth is utilized to neutralize antiseptics and disinfectants as well as to detect organisms remaining after treatment. This broth is especially suited for environmental sampling where neutralization of the chemical is important to determining the bactericidal activity.
  • Bottles are tough and sturdy to reduce breakage
  • Secure tight lid ensures product integrity
  • Bottles are made of clarified polypropylene for clear viewing
  • Bottles have excellent impact and chemical resistance
Caution: This product is not intended to be used for the diagnosis of human disease.