Cable Ties, Solid-Design, Antylia Scientific
Supplier: Antyila Scientific
06830-30
76377-410BG 76.57 USD
76377-410
Cable Ties, Solid-Design, Antylia Scientific
Ties
These ties are injection-molded of 6/6 nylon. They won’t corrode or stretch.
  • Simply bend tip and pull through locking mechanism to secure
  • Won't lose tension, even in vibrating, oily, or wet environments
  • Withstand temperatures to 85 °C (185 °F), with melting point of 257 °C (495 °F)
Durable locking mechanism holds securely even when used to bundle vibrating or flexing materials. Ties continue to hold tension even in oily spray or wet environments. Easily apply ties by hand or use a tensioning tool.
Certifications: UL 94 Flam. class: V-2. Meet military specification MS3367.
Ordering information: Tools have to be ordered separately.