pH Electrode Saver Bottles, Cole-Parmer®
Supplier: Antyila Scientific
05990-99
76377-400EA 23.41 USD
76377-400
pH Electrode Saver Bottles, Cole-Parmer®
Electrode Holders
These storage bottles keep electrodes in working order.
  • Protect against bumps, spills, and drying out
  • Accepts electrodes up to Ø 13 mm
Large bottle includes one solid cap to reduce evaporation of storage solution and one O-ring cap for use with electrodes. Small bottle includes one O-ring cap and is a cost-effective option when storage solution is discarded and not regularly re-used.